Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma Hi-Flow™ Plus membrane 75 Sheets

Catalog No. p-200089062
Click to view available options
Thickness (English):
2 mil
4 mil

Lateral flow test sheets with a nominal capillary flow time of 75 s/4 cm.

  • Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
  • Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
  • Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
  • Contains 5 sample sheets of backed nitrocellulose membrane with a nominal capillary flow time of 75 s/4 cm
  • Dimensions: 30.5 x 25 cm
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Membrane Filter
Format Sheet
Material Nitrocellulose
Sterility Non-sterile
Flow Rate 75 sec./4 cm
Quantity 5
Thickness (English) 2 mil
Type Lateral Flow Membrane
Dimensions (L x W) 30.5 x 25 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.