Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ Hi-Vis Crinkle Latex Coated Seamless Knit Acrylic Gloves

Catalog No. 19046401 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
Medium
Small
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-401 Medium
19-046-402 Large
19-046-403 X-Large
19-046-400 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-046-401 Supplier PIP™ Supplier No. 411420/M
This item is not returnable. View return policy

Textured latex coating enhances grip capabilities in wet or dry conditions, in addition to providing abrasion resistance. Acylic material provides economical cold-weather protection for refrigeration, food processing, and outdoor winter activities.

  • Acrylic material provides economical cold weather protection
  • Latex coating enhances grip capabilities in wet/dry conditions
  • Knit wrist prevents dirt and debris from entering the glove
  • Hi-vis coloring increases worker safety in low-light environments
  • Gloves quickly evaporate moisture and perspiration from the skin
  • Launderable fabric extends glove life and reduce replacement costs

Specifications

Construction Seamless Knit
Coating Material Latex
Color Hi-vis Yellow, Black
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Cold-resistant Gloves
Size Medium
For Use With (Application) Refrigeration, Food Processing, Commercial Fishing, and Construction
Glove Cuff Style Knit
Liner Material Acrylic
Series 41-1420
Yarn Gauge 7
Length (English) 10.7 in.
Length (Metric) 27 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.