Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ PolyKor™ Hi-Vis Seamless Knit HPPE Gloves with Nitrile Foam Coating

Catalog No. 19821997 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-997 Medium
19-821-998 Large
19-821-999 X-Large
19-821-615 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-821-997 Supplier PIP™ Supplier No. 705CGNF/M
Add to Cart
Add to Cart
This item is not returnable. View return policy

HPPE blended shell is lightweight to provide dexterity, tactile sensitivity, and cut resistance. Foam nitrile coating is designed with a cell structure that disperses fluids away on contact for an improved grip.

  • Seamless construction increases comfort and breathability
  • HPPE blended shell is lightweight to provide dexterity, tactile sensitivity, and cut resistance
  • Foam nitrile coating is breathable and durable
  • Coating is designed with a cell structure that disperses fluids away on contact for an improved grip
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Hi-vis coloring makes this product very easy to see

Specifications

ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Nitrile Foam
Color Hi-vis Green, Black
Cut Resistance Rating ANSI A3
Exterior Finish Fully Textured
Hand Specific
Material HPPE
Product Type Seamless Knit Gloves
Size Medium
Style Standard
For Use With (Application) Electronics, Glass Cutting, Sheet Metal Handling, Parts Assembly, and Construction
Glove Cuff Style Knit
Series 705CGNF
Yarn Gauge 10
Length (English) 9.2 in.
Length (Metric) 23.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.