Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ PolyKor™ Hi-Vis Seamless Knit HPPE Gloves with Nitrile Foam Coating

Catalog No. p-7180088
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

HPPE blended shell is lightweight to provide dexterity, tactile sensitivity, and cut resistance. Foam nitrile coating is designed with a cell structure that disperses fluids away on contact for an improved grip.

  • Seamless construction increases comfort and breathability
  • HPPE blended shell is lightweight to provide dexterity, tactile sensitivity, and cut resistance
  • Foam nitrile coating is breathable and durable
  • Coating is designed with a cell structure that disperses fluids away on contact for an improved grip
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Hi-vis coloring makes this product very easy to see

Specifications

ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Nitrile Foam
Color Hi-vis Green, Black
Cut Resistance Rating ANSI A3
Hand Specific
Material HPPE
Product Type Seamless Knit Gloves
Size Medium
For Use With (Application) Electronics, Glass Cutting, Sheet Metal Handling, Parts Assembly, and Construction
Glove Cuff Style Knit
Series 705CGNF
Yarn Gauge 10
Length (English) 9.2 in.
Length (Metric) 23.4 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.