Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567304 Supplier Invitrogen™ Supplier No. PA567304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEHTNLSLRSS SWRKHCGSHS HPHNTQAPGE AKEPLLQDTN Highest antigen sequence identity to the following orthologs - mouse 98%, rat 95%.

Mfsd14a is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIAT1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MFSD14A
Gene Accession No. Q96MC6
Gene Alias Hiat1; hippocampus abundant gene transcript 1; hippocampus abundant transcript 1; hippocampus abundant transcript 1 protein; I79_022140; major facilitator superfamily domain containing 14A; major facilitator superfamily domain-containing 14A; Mfsd14a; NEWGENE_1305281; Putative tetracycline transporter-like protein; tetracycline transporter-like protein
Gene Symbols MFSD14A
Host Species Rabbit
Immunogen Recombinant Human HIAT1. Recombinant protein control fragment (Product #RP-107943).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64645
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.