Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139982
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GKKKKRAANR SIMLAKKIII KDGGTPQGIG SPS.
Mfsd14a is a protein coding gene.
Specifications
| HIAT1 | |
| Polyclonal | |
| Unconjugated | |
| MFSD14A | |
| Hiat1; hippocampus abundant gene transcript 1; hippocampus abundant transcript 1; hippocampus abundant transcript 1 protein; I79_022140; major facilitator superfamily domain containing 14A; major facilitator superfamily domain-containing 14A; Mfsd14a; NEWGENE_1305281; Putative tetracycline transporter-like protein; tetracycline transporter-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64645 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96MC6 | |
| MFSD14A | |
| Recombinant protein corresponding to Human MFSD14A. Recombinant protein control fragment (Product #RP-109321). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction