Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIATL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570475 Supplier Thermo Scientific Supplier No. PA570475
Add to Cart
Add to Cart

HIATL1 Polyclonal Antibody

HIATL1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen HIATL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MFSD14B
Gene Accession No. Q5SR56
Gene Alias 5730414C17Rik; AU045608; AW539692; Hiatl1; hippocampus abundant transcript-like 1; hippocampus abundant transcript-like protein 1; major facilitator superfamily domain containing 14B; major facilitator superfamily domain-containing 14B; MFSD14B; RGD1308377
Gene Symbols MFSD14B
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human HIATL1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84641
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.