Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIATL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983992
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3992 Supplier Bioss Supplier No. BS16504R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MFSD14B gene ontology annotations related to this gene include transmembrane transport.

Specifications

Antigen HIATL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MFSD14B
Gene Accession No. Q5SR56
Gene Alias 5730414C17Rik; AU045608; AW539692; BOS_85; Hiatl1; hippocampus abundant transcript-like 1; hippocampus abundant transcript-like 1-like; Hippocampus abundant transcript-like protein 1; LOC100350305; major facilitator superfamily domain containing 14B; Major facilitator superfamily domain-containing 14B; Mfsd14b; RGD1308377
Gene Symbols MFSD14B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from HIATL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84641
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.