Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIBADH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532070 Supplier Invitrogen™ Supplier No. PA532070
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, Neuro2A, C8D30, PC-12. Predicted reactivity: Mouse (92%), Rat (91%), Xenopus laevis (86%), Chicken (87%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

3-hydroxyisobutyrate dehydrogenase (3-hydroxy-2-methylpropanoate:NAD(+) oxidoreductase, EC 1.1. 1.31) is a dimeric mitochondrial enzyme that catalyzes the NAD(+)-dependent, reversible oxidation of 3-hydroxyisobutyrate, an intermediate of valine catabolism, to methylmalonate semialdehyde.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIBADH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene HIBADH
Gene Accession No. P29266, P31937, Q99L13
Gene Alias 3'-hydroxyisobutyrate dehydrogenase; 3-hydroxyisobutyrate dehydrogenase; 3-hydroxyisobutyrate dehydrogenase, mitochondrial; 6430402H10Rik; AI265272; Hibadh; NS5ATP1
Gene Symbols HIBADH
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HIBADH. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11112, 58875, 63938
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.