Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIBADH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554303
Description
Immunogen sequence: NPVPGVMDGV PSANNYQGGF GTTLMAKDLG LAQDSATSTK SPILLGSLAH QIYRMMCAKG YSKKDFSSVF QFLR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
3-hydroxyisobutyrate dehydrogenase (3-hydroxy-2-methylpropanoate:NAD(+) oxidoreductase, EC 1.1. 1.31) is a dimeric mitochondrial enzyme that catalyzes the NAD(+)-dependent, reversible oxidation of 3-hydroxyisobutyrate, an intermediate of valine catabolism, to methylmalonate semialdehyde.
Specifications
HIBADH | |
Polyclonal | |
Unconjugated | |
HIBADH | |
3'-hydroxyisobutyrate dehydrogenase; 3-hydroxyisobutyrate dehydrogenase; 3-hydroxyisobutyrate dehydrogenase, mitochondrial; 6430402H10Rik; AI265272; Hibadh; NS5ATP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11112 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P31937 | |
HIBADH | |
Recombinant protein corresponding to Human HIBADH. Recombinant protein control fragment (Product #RP-92655). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction