Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 100-4.6
Catalog No. M1500137004 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
60 SI
HPLC phase
NH2
RP18
Mobile Phase Compatibility:
NP (normal-phase)
RP (reversed-phase)
RP (when in mode) and NP
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase Mobile Phase Compatibility
M1500137004 HPLC phase RP (reversed-phase)
M1500137052 RP18 RP (reversed-phase)
M1500137076 NH2 RP (when in mode) and NP
M1500137109 60 SI NP (normal-phase)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1500137004 Supplier MilliporeSigma™ Supplier No. 1500137004
Add to Cart
Add to Cart
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma Hibar™ HPLC Columns, 100-4.6 come with customized packing (please specify packing material) and are 100mmL x 4.6mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 4.6 mm
Length (Metric) 100 mm
Mobile Phase Compatibility RP (reversed-phase)
Stationary Phase HPLC phase
Packing Material Customized (please specify packing material)
Column Type Hibar™ HPLC Column