Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 250-3
Catalog No. M1004237079 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
RP18
RP8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase
M1004237079 RP18
M1004237087 RP8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1004237079 Supplier MilliporeSigma™ Supplier No. 1004237079
Only null left
Add to Cart
Add to Cart
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma Hibar™ HPLC Columns, 250-3 come with customized packing (please specify packing material) and are 250mmL x 3mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 3 mm
Length (Metric) 250 mm
Mobile Phase Compatibility RP (reversed-phase)
Stationary Phase RP18
Packing Material Customized (specify packing material)
Column Type Hibar™ HPLC Column