Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIBCH Antibody (OTI3H5) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271796
Description
HIBCH Monoclonal specifically detects HIBCH in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
HIBCH | |
Monoclonal | |
LYOPH | |
Western Blot 1:1000-2000, Flow Cytometry 1:100, Immunohistochemistry 1:50, CyTOF-ready | |
3-hydroxyisobutyryl-CoA hydrolase, EC 3.1.2.4,3-hydroxyisobutyryl-coenzyme A hydrolase3-hydroxyisobutyryl-CoA hydrolase, mitochondrial, HIB-CoA hydrolase, HIBYL-CoA-H, HIBYLCOAH | |
Mouse | |
39.4 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
OTI3H5 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
HIBCH | |
Full length human recombinant protein of human HIBCH (NP_055177) produced in HEK293T cell. | |
Affinity Purified | |
Lipid and Metabolism | |
26275 | |
Human, Canine | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction