Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIBCH Antibody (OTI3H5), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271796UV
Description
HIBCH Monoclonal specifically detects HIBCH in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
HIBCH | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HIBCH | |
Full length recombinant protein of human HIBCH (NP_055177) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Canine | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI3H5 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
3-hydroxyisobutyryl-CoA hydrolase, EC 3.1.2.4,3-hydroxyisobutyryl-coenzyme A hydrolase3-hydroxyisobutyryl-CoA hydrolase, mitochondrial, HIB-CoA hydrolase, HIBYL-CoA-H, HIBYLCOAH | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
26275.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction