Learn More
HIBCH Mouse anti-Human, Clone: OTI2A5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501318

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Beta-hydroxyisobutyryl-CoA hydrolase (EC 3.1.2.4) is responsible for the specific hydrolysis of HIBYL-CoA, a valine catabolite, as well as the hydrolysis of beta-hydroxypropionyl-CoA, an intermediate in a minor pathway of propionate metabolism.Specifications
HIBCH | |
Monoclonal | |
Unconjugated | |
HIBCH | |
HIBYLCOAH | |
Mouse | |
Affinity Chromatography | |
RUO | |
26275 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2A5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q6NVY1 | |
HIBCH | |
Full length human recombit protein of human HIBCH produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.