Learn More
Invitrogen™ HIC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589523
Description
Immunogen sequence: AATPVIQACY PSPVGPPPPP AAEPPSGPEA AVNTHCAELY ASGPGPAAAL CASERRCSPL CGLDLSKKSP PGSAAPERPL AERELPPRPD SPPSAGPAAY KEPPLALPSL PPLPFQKLEE AAPPSDPFRG GSGSPGPEPP GRPDGPSLLY RWMKHEPGLG SYGDELGRER GSPSERCEER GGDAAVSPGG PPLGLAPPPR YPGSLDGPGA GGDGDDYKSS SEETGSSEDP SPPGGHLEGY PCPHLAYGEP E; Positive Samples: Mouse liver; Cellular Location: Nucleus.
Hypermethylated in cancer (HIC-1) was originally identified as a target of p53-induced gene expression. HIC-1 is deleted in the genetic disorder Miller-Dieker syndrome (MDS), and the expression of HIC-1 is also frequently suppressed in leukemia and various cancers due to the hypermethylation of specific DNA regions and the resulting transcriptional silencing. These and other studies indicate that HIC-1 acts as a putative tumor suppressor protein that mediates transcriptional repression. HIC-1 is ubiquitously expressed in adult tissues and its structure is defined by five zinc fingers and an N-terminal broad complex POZ (or BTB) domain. In several BTB/POZ containing proteins, including BCL-6 and the promyelocytic leukemia zinc-finger (PLZF) oncoprotein, this domain interacts with the SMRT/N-CoR-mSin3A HDAC complex and is directly involved in repressing and silencing gene transcription. When this domain is deleted, as with the oncogenic PLZF-RAR chimera of promyelocytic leukemias, this transcriptional repression is attenuated. Conversely, HIC-1 does not interact with components of the HDAC complex, suggesting that HIC-1-induced transcriptional repression is unassociated with the POZ/BTB domain.
Specifications
HIC1 | |
Polyclonal | |
Unconjugated | |
Hic1 | |
AA408311; HIC1; hic-1; hypermethylated in cancer 1; hypermethylated in cancer 1 protein; ZBTB29; Zinc finger and BTB domain-containing protein 29; ZNF901 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15248 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.6 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9R1Y5 | |
Hic1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 180-430 of human HIC1 (NP_0010916721). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.