Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597235 Supplier Invitrogen™ Supplier No. PA597235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AATPVIQACY PSPVGPPPPP AAEPPSGPEA AVNTHCAELY ASGPGPAAAL CASERRCSPL CGLDLSKKSP PGSAAPERPL AERELPPRPD SPPSAGPAAY KEPPLALPSL PPLPFQKLEE AAPPSDPFRG GSGSPGPEPP GRPDGPSLLY RWMKHEPGLG SYGDELGRER GSPSERCEER GGDAAVSPGG PPLGLAPPPR YPGSLDGPGA GGDGDDYKSS SEETGSSEDP SPPGGHLEGY PCPHLAYGEP E; Positive Samples: H460, HT-29, DU145, Rat heart; Cellular Location: Nucleus.

Hypermethylated in cancer (HIC-1) was originally identified as a target of p53-induced gene expression. HIC-1 is deleted in the genetic disorder Miller-Dieker syndrome (MDS), and the expression of HIC-1 is also frequently suppressed in leukemia and various cancers due to the hypermethylation of specific DNA regions and the resulting transcriptional silencing. These and other studies indicate that HIC-1 acts as a putative tumor suppressor protein that mediates transcriptional repression. HIC-1 is ubiquitously expressed in adult tissues and its structure is defined by five zinc fingers and an N-terminal broad complex POZ (or BTB) domain. In several BTB/POZ containing proteins, including BCL-6 and the promyelocytic leukemia zinc-finger (PLZF) oncoprotein, this domain interacts with the SMRT/N-CoR-mSin3A HDAC complex and is directly involved in repressing and silencing gene transcription. When this domain is deleted, as with the oncogenic PLZF-RAR chimera of promyelocytic leukemias, this transcriptional repression is attenuated. Conversely, HIC-1 does not interact with components of the HDAC complex, suggesting that HIC-1-induced transcriptional repression is unassociated with the POZ/BTB domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Hic1
Gene Accession No. Q14526
Gene Alias AA408311; HIC1; hic-1; hypermethylated in cancer 1; hypermethylated in cancer 1 protein; ZBTB29; Zinc finger and BTB domain-containing protein 29; ZNF901
Gene Symbols Hic1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-430 of human HIC1 (NP_0010916721).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303310, 3090
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.