Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIC2 Antibody (PCRP-HIC2-2F8), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314235MFV500
Description
HIC2 Monoclonal antibody specifically detects HIC2 in Human samples. It is validated for Flow CytometrySpecifications
HIC2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
23119 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
PCRP-HIC2-2F8 | |
Flow Cytometry | |
HIC1-related gene on chromosome 22 protein, Hic-2, Hic-3, HRG22KIAA1020hic-2, hypermethylated in cancer 2, hypermethylated in cancer 2 protein, ZBTB30hic-3, Zinc finger and BTB domain-containing protein 30, ZNF907 | |
Recombinant full-length human HIC2protein (Uniprot: Q96JB3) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction