Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIC2 Polyclonal Antibody
HIC2 Polyclonal Antibody
Supplier: Thermo Scientific PA5103138
Description
HIC2 Polyclonal Antibody for Western Blot

Specifications
HIC2 | |
Polyclonal | |
Unconjugated | |
HIC2 | |
AA409108; HIC ZBTB transcriptional repressor 2; HIC1-related gene on chromosome 22 protein; HIC2; hic-2; Hic-3; HRG22; hypermethylated in cancer 2; hypermethylated in cancer 2 protein; Kiaa1020; LOW QUALITY PROTEIN: hypermethylated in cancer 2 protein; mKIAA1020; ZBTB30; Zinc finger and BTB domain-containing protein 30; ZNF907 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23119, 58180 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96JB3, Q9JLZ6 | |
HIC2 | |
A synthesized peptide derived from human HIC2(Accession Q96JB3), corresponding to amino acid residues A168-V218. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction