Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540430
Description
Peptide sequence: CKEEEENGKD ASEDSAQSGS EGGSGHASAH YMYRQEGYET VSYGDNLYVC Sequence homology: Cow: 85%; Dog: 85%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 85%; Rabbit: 77%; Rat: 85%.
HIC2 contains 5 C2H2-type zinc fingers and 1 BTB (POZ) domain. It belongs to the krueppel C2H2-type zinc-finger protein family, Hic subfamily and is a transcriptional repressor.
Specifications
HIC2 | |
Polyclonal | |
Unconjugated | |
HIC2 | |
AA409108; HIC ZBTB transcriptional repressor 2; HIC1-related gene on chromosome 22 protein; HIC2; hic-2; Hic-3; HRG22; hypermethylated in cancer 2; hypermethylated in cancer 2 protein; Kiaa1020; LOW QUALITY PROTEIN: hypermethylated in cancer 2 protein; mKIAA1020; ZBTB30; Zinc finger and BTB domain-containing protein 30; ZNF907 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23119 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96JB3 | |
HIC2 | |
Synthetic peptide directed towards the middle region of human HIC2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction