Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111031
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111031 100 μL
PIPA5111485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111031 Supplier Invitrogen™ Supplier No. PA5111031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIQARYQGLV DGRKGAHAPQ ELPQAKGSDD ELFLGGSNQD SVQGLGRAVC PAGGEAGLGG CSSSTNGSSG GCEQELGLD Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 81%.

HIC2 contains 5 C2H2-type zinc fingers and 1 BTB (POZ) domain. It belongs to the krueppel C2H2-type zinc-finger protein family, Hic subfamily and is a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HIC2
Gene Accession No. Q96JB3
Gene Alias AA409108; HIC ZBTB transcriptional repressor 2; HIC1-related gene on chromosome 22 protein; HIC2; hic-2; Hic-3; HRG22; hypermethylated in cancer 2; hypermethylated in cancer 2 protein; Kiaa1020; LOW QUALITY PROTEIN: hypermethylated in cancer 2 protein; mKIAA1020; ZBTB30; Zinc finger and BTB domain-containing protein 30; ZNF907
Gene Symbols HIC2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human HIC2. Recombinant protein control fragment (Product #RP-95370).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23119
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.