Learn More
Invitrogen™ HIC2 Polyclonal Antibody

Description
Immunogen sequence: VIQARYQGLV DGRKGAHAPQ ELPQAKGSDD ELFLGGSNQD SVQGLGRAVC PAGGEAGLGG CSSSTNGSSG GCEQELGLD Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 81%.
Specifications
Specifications
| Antigen | HIC2 |
| Applications | ChIP Assay, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HIC2 |
| Gene Accession No. | Q96JB3 |
| Gene Alias | AA409108; HIC ZBTB transcriptional repressor 2; HIC1-related gene on chromosome 22 protein; HIC2; hic-2; Hic-3; HRG22; hypermethylated in cancer 2; hypermethylated in cancer 2 protein; Kiaa1020; LOW QUALITY PROTEIN: hypermethylated in cancer 2 protein; mKIAA1020; ZBTB30; Zinc finger and BTB domain-containing protein 30; ZNF907 |
| Gene Symbols | HIC2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.