Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528839 Supplier Invitrogen™ Supplier No. PA528839
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Hic-5 tansfected 293T cell. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a coactivator of the androgen receptor, a transcription factor which is activated by androgen and has a key role in male sexual differentiation. The encoded protein is thought to regulate androgen receptor activity and may have a role to play in the treatment of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIC5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Tgfb1i1
Gene Accession No. O43294
Gene Alias Androgen receptor coactivator 55 kDa protein; androgen receptor coactivator ARA55; androgen receptor-associated protein of 55 kDa; Ara55; Hic5; hic-5; hydrogen peroxide-inducible clone 5 protein; hydrogen peroxide-inducible clone-5; TGF beta-stimulated clone 5; Tgfb1i1; transforming growth factor beta 1 induced transcript 1; transforming growth factor beta-1-induced transcript 1 protein; TSC-5
Gene Symbols Tgfb1i1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human HIC5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7041
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.