Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117133
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117133 Supplier Invitrogen™ Supplier No. PA5117133
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HT-1080, Mouse spleen Immunogen sequence: VASGEQKEDQ SEDKKRPSLP SSPSPGLPKA SATSATLELD RLMASLSDFR VQNHLPASGP TQPPVVSSTN EGSPSPPEPT GKGSLDTMLG LLQSDLSRRG V.

This gene encodes a coactivator of the androgen receptor, a transcription factor which is activated by androgen and has a key role in male sexual differentiation. The encoded protein is thought to regulate androgen receptor activity and may have a role to play in the treatment of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIC5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Tgfb1i1
Gene Accession No. O43294, Q62219, Q99PD6
Gene Alias Androgen receptor coactivator 55 kDa protein; androgen receptor coactivator ARA55; androgen receptor-associated protein of 55 kDa; Ara55; Hic5; hic-5; hydrogen peroxide-inducible clone 5 protein; hydrogen peroxide-inducible clone-5; TGF beta-stimulated clone 5; Tgfb1i1; transforming growth factor beta 1 induced transcript 1; transforming growth factor beta-1-induced transcript 1 protein; TSC-5
Gene Symbols Tgfb1i1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-220 of human TGFB1I1 (NP_0010359191).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21804, 7041, 84574
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.