Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HICE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596830 Supplier Invitrogen™ Supplier No. PA596830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLAEFERRAE KNLLIMCKEK EKLQKKAHEL KRRLLLSQRK RELADVLDAQ IEMLSPFEAV ATRFKEQYRT FATALDTTRH ELPVRSIHLE GDGQQLLDAL QHELVTTQRL LGELDVGDSE ENVQVLDLLS ELKDVTAKKD LELRRSFAQV LELSAEASKE AALANQEVWE ETQGMAPPSR WYFNQDSACR ESGGAPKNTP LSEDDNPGAS SAPAQATFIS PSEDFSSSSQ AEVPPSLSRS GRDLS; Positive Samples: Jurkat, NCI-H460, Mouse testis; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, spindle, spindle pole.

Hice1 contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HICE1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HAUS8
Gene Accession No. Q99L00, Q9BT25
Gene Alias 2410004L22Rik; 4HAUS augmin-like complex, subunit 8; DGT4; HAUS augmin like complex subunit 8; HAUS augmin-like complex subunit 8; HAUS augmin-like complex, subunit 8; HAUS8; HEC1/NDC80 interacting, centrosome associated 1; HEC1/NDC80-interacting centrosome-associated protein 1; Hec1-interacting and centrosome-associated 1; Hice1; MGC20533; NY SAR 48; NYSAR48; NY-SAR-48; Sarcoma antigen NY-SAR-48; sarcoma antigen NY-SAR-48 homolog
Gene Symbols HAUS8
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76478, 93323
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.