Learn More
Synthware™ Hickman-Hinkle Distillation Head with Side Port
Shop All Kemtech Products
Click to view available options
Standard Taper Joint:
14/10
Description
This head has a threaded side port with septum and cap for easier syringe access to well contents. Supplied with one compression cap, one Viton™ O-ring and one PTFE-faced silicone rubber septum
Specifications
Specifications
| Includes | Compression Cap, O-Ring, Rubber Septum |
| For Use With (Application) | Organic Synthesis |
| Type | Hickman-Hickman-Hinkle Still Head |
| Material | Borosilicate Glass |
| Quantity | 1 Ea. |
| Standard Taper Joint | 14/10 |
| Autoclavable | Autoclavable |
| Size | 14/10 |