Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HID1 Monoclonal Antibody (OTI2F4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525149

Catalog No. PIMA525149


Only null left
Add to Cart

Description

Description

HID1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

May play an important role in the development of cancers in a broad range of tissues. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HID1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IV36
HID1
Full length human recombinant protein of C17orf28 produced in HEK293T cell.
100 μL
Primary
Human, Rat, Canine, Monkey
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI2F4
Unconjugated
HID1
C17orf28; C630004H02Rik; C9H17orf28; DMC1; downregulated in multiple cancer 1; Down-regulated in multiple cancers 1; Hid1; HID-1; HID1 domain containing; HID1 domain-containing protein; protein HID1; Protein hid-1 homolog; RGD1311422; UPF0663 transmembrane protein C17orf28; UPF0663 transmembrane protein C17orf28 homolog
Mouse
Affinity chromatography
RUO
283987, 287822, 483305
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.