Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HID1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565096 Supplier Invitrogen™ Supplier No. PA565096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALPLFTSLLNT VCAYDPVGYG IPYNHLLFSD YREPLVEEAA QVLIVTLDHD SASSASPTVD GTTTGTAMDD ADPPGPENLF VNYLSRIHRE EDFQFILKGI ARLLSNPLLQ TYLPNSTKKI QFHQELLVLF WKLCDFNKKF L Highest antigen sequence identity to the following orthologs - mouse 99%, rat 98%.

May play an important role in the development of cancers in a broad range of tissues. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HID1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HID1
Gene Accession No. Q8IV36
Gene Alias C17orf28; C630004H02Rik; C9H17orf28; DMC1; downregulated in multiple cancer 1; Down-regulated in multiple cancers 1; Hid1; HID-1; HID1 domain containing; HID1 domain-containing protein; protein HID1; Protein hid-1 homolog; RGD1311422; UPF0663 transmembrane protein C17orf28; UPF0663 transmembrane protein C17orf28 homolog
Gene Symbols HID1
Host Species Rabbit
Immunogen Recombinant Human HID1. Recombinant protein control fragment (Product #RP-104630).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283987
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.