Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HID1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100412 Supplier Invitrogen™ Supplier No. PA5100412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C17orf28.

May play an important role in the development of cancers in a broad range of tissues. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HID1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HID1
Gene Accession No. Q8IV36, Q8R1F6
Gene Alias C17orf28; C630004H02Rik; C9H17orf28; DMC1; downregulated in multiple cancer 1; Down-regulated in multiple cancers 1; Hid1; HID-1; HID1 domain containing; HID1 domain-containing protein; protein HID1; Protein hid-1 homolog; RGD1311422; UPF0663 transmembrane protein C17orf28; UPF0663 transmembrane protein C17orf28 homolog
Gene Symbols HID1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HID1(Accession Q8IV36), corresponding to amino acid residues E283-H333.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217310, 283987, 287822
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.