Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIDE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571557
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571557 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571557 Supplier Invitrogen™ Supplier No. PA571557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIDE1 Polyclonal Antibody for Western Blot

HIDE1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIDE1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene C19orf38
Gene Accession No. A8MVS5
Gene Alias C19orf38; chromosome 19 open reading frame 38; HIDE1; Highly expressed in immature dendritic cell transcript 1; protein HIDE1
Gene Symbols C19orf38
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 129-158 amino acids from the Central region of human C19orf38.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.