Learn More
Description
Specifications
Specifications
| Antigen | HIF-1 alpha |
| Applications | ChIP Assay, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Gel Shift, ChIP Assay, Immunoassay |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, Simple Western 1:100, ELISA, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:100 - 1:500, Immunoblotting, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Chromatin Immunoprecipitation Sequencing reported in scientific literature (PMID 34277635), Dual RNAscope ISH-IHC |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q61221, O35800 |
| Gene Alias | AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp |
| Gene Symbols | HIF1A |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
