Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIF-1 alpha Antibody, DyLight 488 - Exon 13, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP147181G
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot.Specifications
HIF-1 alpha | |
Polyclonal | |
Western Blot | |
Q16665 | |
HIF1A | |
This HIF-1 alpha Antibody - Exon 13 was developed against a synthetic peptide made to an internal portion of the human HIF-1 alpha protein (within amino acids 700 - 750) [Uniprot# Q16665]. | |
Affinity Purified | |
RUO | |
Primary | |
HIF-1 alpha Antibody - Exon 13 is specific for Exon 13 of HIF-1 alpha. | |
Store at 4C in the dark. |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
Rabbit | |
93 kDa | |
0.1 mL | |
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
3091 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction