Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIF-1 alpha Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100479R
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Porcine, Canine, Fish, Goat, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount.Specifications
HIF-1 alpha | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
Rabbit | |
93 kDa | |
0.1 mL | |
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
3091 | |
Human, Mouse, Rat, Pig, Canine, Fish, Goat, Hamster, Primate, Rabbit | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HIF1A | |
This HIF-1 alpha Antibody was developed against a fusion protein including amino acids 530 - 825 of the mouse HIF-1 alpha protein [Uniprot# Q61221]. | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, rat, goat, primate and rabbit. Hamster reactivity reported in scientific literature (PMID: 24997364 and 24997360). | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction