Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIF-1 alpha Antibody (H1alpha 67-7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | HIF-1 alpha |
---|---|
Clone | H1alpha 67-7 |
Concentration | 1.5 mg/mL |
Dilution | Western Blot 1:500 - 1:1000 |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100130SS
![]() |
Novus Biologicals
NB100130SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB100130
![]() |
Novus Biologicals
NB100130 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human, Monkey samples. It is validated for Western Blot.Specifications
HIF-1 alpha | |
1.5 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
Q16665 | |
3091 | |
This HIF-1 alpha Antibody (H1alpha 67-7) was developed against a fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [Uniprot# Q16665]. | |
Primary | |
Store at 4C. Do not freeze. |
H1alpha 67-7 | |
Western Blot 1:500 - 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Monkey | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
HIF1A | |
IgG2b | |
Protein G purified | |
93 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title