Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 35 publications

$293.50 - $611.00

Specifications

Antigen Fibronectin
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported by customer review
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP191258SS
SDP
View Documents
Novus Biologicals
NBP191258SS
0.025 mL
Each for $293.50
Only null left
Add to Cart
 
NBP191258
SDP
View Documents
Novus Biologicals
NBP191258
0.1 mL
Each for $611.00
Only null left
Add to Cart
 
Description

Description

Fibronectin Polyclonal specifically detects Fibronectin in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Fibronectin
Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported by customer review
Polyclonal
Rabbit
Cancer, Cellular Markers, Mesenchymal Stem Cell Markers, Stem Cell Markers
PBS, 30% Glycerol with 0.05% Sodium Azide
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
FN1
IgG
Affinity Purified
Human and Mouse. Based on immunogen's sequence homology, this antibody is predicted to react with Chicken (91%), Rat (95%), Canine/Dog (100%), Bovine (100%) and Equine/Horse (100%). Immunogen similarity with other species: Xenopus (86%) and Newt (82%).
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline
P02751
2335
A synthetic peptide made toward the C-terminal region of the human Fibronectin protein (within residues 2250-2300). [Swiss-Prot: P02751]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.