Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIF-1 alpha Antibody, PE/Cy5.5, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100479PECY55
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat, Porcine, Canine, Fish, Goat, Hamster, Primate, Rabbit samples. It is validated for Flow Cytometry.Specifications
HIF-1 alpha | |
Polyclonal | |
Flow Cytometry | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp | |
Rabbit | |
93 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Pig, Canine, Fish, Goat, Hamster, Primate, Rabbit | |
IgG |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
HIF1A | |
This HIF-1 alpha Antibody was developed against a fusion protein including amino acids 530 - 825 of the mouse HIF-1 alpha protein [Uniprot# Q61221]. | |
Affinity Purified | |
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
3091.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction