Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha (HIF1A) Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
SDP

Catalog No. 502543924
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3924 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3924 Supplier OriGene Supplier No. CF811847
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIF-1 alpha (HIF1A) Monoclonal antibody specifically detects HIF-1 alpha (HIF1A) in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen HIF-1 alpha (HIF1A)
Applications Western Blot
Classification Monoclonal
Clone OTI2D5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Mlh1
Gene Accession No. P40692, P97679, Q9JK91
Gene Alias AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L homeolog; hypoxia-inducible factor 1-alpha; hypoxia-inducible factor-1 alpha; hypoxia-inducible factor1alpha; member of PAS protein 1; member of PAS superfamily 1; MOP1; PAS domain-containing protein 8; PASD8; XELAEV_18039792mg; XELAEV_18041137mg; xhif1a
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-298 of human MLH1 (NP_000240) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17350, 4292, 81685
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.