Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HIF-1 alpha Knockout CoCl2-treated/untreated HeLa Cell Lysate
SDP

Catalog No. NB151154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151154 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151154 Supplier Novus Biologicals™ Supplier No. NBP311826
Only null left
Add to Cart
Add to Cart

HIF-1 alpha KO HeLa Whole cell lysate is provided as a Western blot positive control for Hif-1 research. The lysates are provided as a set of treated and untreated samples. Use 10 ul per lane for a standard mini-gel blot (approx. 1 mg/ml).

Specifications

For Use With (Application) SDS-Page
Quantity 2 Vials
Storage Buffer The protein lysates are prepared in 1 x Laemmli sample buffer with BME. Boiling before loading is not necessary.
Concentration 1.0 mg/ml
Target HIF-1 alpha
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.