Learn More
HIF-1 alpha Monoclonal Antibody (Mgc3), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$436.00
Specifications
Antigen | HIF-1 alpha |
---|---|
Clone | Mgc3 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
17-752-882
![]() |
Invitrogen™
17752882 |
100 μg |
Each of 1 for $436.00
|
![]() |
|||||
Description
Description: This Mgc3 monoclonal antibody detects hypoxia-inducible factor 1 alpha (HIF-1 alpha) from human, non-human primate, bovine, mouse and porcine cells. This antibody does not cross-react with ARNT or the related HIF-2 alpha. Mgc3 has been successfully used in western blotting, immunofluorescence, immunocytochemistry, immunoprecipitation, flow cytometry and gel shift procedures. By western blot, this antibody detects a ∽93 kDa protein representing HIF-1 alpha after hypoxic induction in COS cells. Immunofluorescence staining of HIF-1 alpha in COS-7 cells with clone Mgc3 yields nuclear staining after exposing cells to 1% oxygen for 4 hours. In gel shift assay experiments, the Mgc3 antibody detected only mouse and not human HIF-1 alpha. Other anti-HIF1 alpha antibodies were previously included in a Thermo Scientific Cellomics High Content Screening Kit. Clone Mgc3 is now recommended as a replacement. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. High content assays may require further optimization, including the selection of the optimal Dylight-conjugated secondary antibody. Applications Reported: This Mgc3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This Mgc3 antibody has been tested by flow cytometric analysis of DFOA stimulated HeLa cells using the methanol fixation protocol. This may be used at less than or equal to 1.0 μg/mL per test.
HIF1-alpha (HIF1A) is a subunit of HIF1, which is a transcription factor found in mammalian cells cultured under reduced oxygen tension. HIF-1 is a heterodimer consisting of an alpha and beta subunit, both belonging to the basic-helix-loop-helix Per-aryl hydrocarbon receptor nuclear translocator-Sim (PAS) family of transcription factors. HIF1 functions as a transcriptional regulator of the adaptive response to hypoxia. Under hypoxic conditions, HIF-1 activates the transcription of over 40 genes, including erythropoietin, glucose transporters, glycolytic enzymes, vascular endothelial growth factor, HILPDA, and other genes whose protein products increase oxygen delivery or facilitate metabolic adaptation to hypoxia. HIF1-alpha regulates hypoxia-mediated apoptosis, cell proliferation and tumor angiogenesis. Hypoxia which induces p53 protein accumulation, directly interacts with HIF1-alpha and reduces hypoxia-induced expression of HIF1-alpha by promoting MDM2-mediated ubiquitination and proteasomal degradation under hypoxic conditions. Recent studies suggest that induction of NOX4 by HIF1-alpha contributes to maintain ROS levels after hypoxia and hypoxia-induced proliferation. In humans, it is located on the q arm of chromosome 14. The C-terminal of HIF1A binds to p300. p300/CBP-HIF complexes participate in the induction of hypoxia-responsive genes, including VEGF. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease.Specifications
HIF-1 alpha | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q16665, Q61221 | |
HIF1A | |
IgG1 κ | |
4°C, store in dark, DO NOT FREEZE! | |
HIF1A |
Mgc3 | |
Flow Cytometry | |
APC | |
Mouse | |
Human, Mouse | |
15251, 3091 | |
Human HIF-1 alpha amino acids 530-826. | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.