Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIF-1 beta Monoclonal Antibody (H1beta234), DyLight™ 488
GREENER_CHOICE

Catalog No. PIMA516013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516013 Supplier Invitrogen™ Supplier No. MA516013
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIF-1 beta Monoclonal Antibody for ChIP, EMSA, ICC/IF, IHC (P), IP, Western Blot

HIF-1 beta is a series of aryl hydrocarbon receptor nuclear translocator (ARNT) gene products. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease. HIF-1 is a nuclear protein involved in mammalian oxygen homeostasis. This occurs as a posttranslational modification by prolyl hydroxylation. HIF-1 is a heterodimer composed of HIF-1 alpha and HIF-1 beta subunits. Both subunits are constantly translated. However, under normoxic conditions, human HIF-1 alpha is hydroxylated at Pro402 or Pro564 by a set of HIF prolyl hydroxylases, is polyubiquinated, and eventually degraded in proteosomes. Under hypoxic conditions, the lack of hydroxylation prevents HIF degradation and increases transcriptional activity. Therefore, the concentration of HIF-1 alpha increases in the cell. In contrast, HIF-1 beta remains stable under either condition. HIF-1 beta is a series of aryl hydrocarbon receptor nuclear translocator (ARNT) gene products. Diseases associated with HIF-1 beta dysfunction include hypoxia and renal cell carcinoma.HIF-1 beta is a series of aryl hydrocarbon receptor nuclear translocator (ARNT) gene products. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease. HIF-1 is a nuclear protein involved in mammalian oxygen homeostasis. This occurs as a posttranslational modification by prolyl hydroxylation. HIF-1 is a heterodimer composed of HIF-1 alpha and HIF-1 beta subunits. Both subunits are constantly translated. However, under normoxic conditions, human HIF-1 alpha is hydroxylated at Pro402 or Pro564 by a set of HIF prolyl hydroxylases, is polyubiquinated, and eventually degraded in proteosomes. Under hypoxic conditions, the lack of hydroxylation prevents HIF degradation and increases transcriptional activity. Therefore, the concentration of HIF-1 alpha increases in the cell. In contrast, HIF-1 beta remains stable under either condition. HIF-1 beta is a series of aryl hydrocarbon receptor nuclear translocator (ARNT) gene products. Diseases associated with HIF-1 beta dysfunction include hypoxia and renal cell carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF-1 beta
Applications ChIP Assay, Gel Shift, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone H1beta234
Concentration 0.70 mg/mL
Conjugate DyLight 488
Formulation 50mM sodium borate with 0.05% sodium azide
Gene ARNT
Gene Accession No. P27540, P41739, P53762, Q9BE97
Gene Alias AHA-1; ARNT; ARNT protein; Arnt1; ARNT1a; ARNT1b; aryl hydrocarbon receptor nuclear translocater; Aryl hydrocarbon receptor nuclear translocator; Aryl hydrocarbon receptor nuclear translocator 1; aryl hydrocarbon receptor nuclear translocator type 1a; aryl hydrocarbon receptor nuclear translocator type 1b; bHLHe2; Class E basic helix-loop-helix protein 2; D3Ertd557e; dioxin receptor, nuclear translocator; Drnt; ESTM42; hif 1; HIF1 beta; HIF-1 beta; Hif1b; HIF1BETA; HIF-1beta; HIF-1-beta; HIF1-beta; hypoxia-inducible factor 1 beta; hypoxia-inducible factor 1, beta subunit; hypoxia-inducible factor 1-beta; mKIAA4051; TANGO; W08714; zgc:136664
Gene Symbols ARNT
Host Species Mouse
Immunogen Fusion protein containing amino acids 496-789 of human HIF-1 beta.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11863, 25242, 281010, 405, 443285
Target Species Human, Mouse, Rat, Monkey, Bovine, Sheep, Ferret
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.