Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIF-2 alpha Polyclonal Antibody, DyLight™ 488
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522695
Description
Suggested positive control: hypoxic lysates.
HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.
Specifications
HIF-2 alpha | |
Polyclonal | |
DyLight 488 | |
EPAS1 | |
basic-helix-loop-helix-PAS protein MOP2; bHLHe73; Class E basic helix-loop-helix protein 73; ECYT4; endothelial PAS domain protein 1; endothelial PAS domain protein 1b; endothelial PAS domain-containing protein 1; endothelial PAS domain-containing protein 1b; EPAS; Epas1; EPAS-1; epas1b; Hif like protein; HIF1 alpha-like factor; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF1alpha-like factor; HIF2 alpha; HIF-2 alpha; HIF2A; hif-2a; hif-2ab; HIF-2alpha; HIF-2-alpha; HIF2-alpha; HIF-related factor; HLF; HLF (HIF1alpha-like factor); HRF; hypoxia inducible factor 2 alpha; hypoxia inducible factor 2, alpha subunit; hypoxia inducible factor 2a; hypoxia inducible transcription factor 2alpha; hypoxia-inducible factor 2 alpha; hypoxia-inducible factor 2-alpha; Member of PAS protein 2; mHLF; MOP2; PAS domain-containing protein 2; PASD2; wu:fq36f01 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100348809, 100771160, 13819, 2034, 29452, 443286 | |
4°C, store in dark | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.83 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
P97481, Q99814, Q9JHS1 | |
EPAS1 | |
A peptide derived from the C-terminus of mouse/human HIF-2 alpha protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Rabbit, Monkey, Sheep, Hamster, Fish, Reptile | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction