Learn More
Invitrogen™ HIF-2 alpha Polyclonal Antibody, DyLight™ 650

Description
Suggested positive control: hypoxic lysates.
Specifications
Specifications
| Antigen | HIF-2 alpha |
| Applications | ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.64 mg/mL |
| Conjugate | DyLight 650 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | EPAS1 |
| Gene Accession No. | P97481, Q99814, Q9JHS1 |
| Gene Alias | basic-helix-loop-helix-PAS protein MOP2; bHLHe73; Class E basic helix-loop-helix protein 73; ECYT4; endothelial PAS domain protein 1; endothelial PAS domain protein 1b; endothelial PAS domain-containing protein 1; endothelial PAS domain-containing protein 1b; EPAS; Epas1; EPAS-1; epas1b; Hif like protein; HIF1 alpha-like factor; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF1alpha-like factor; HIF2 alpha; HIF-2 alpha; HIF2A; hif-2a; hif-2ab; HIF-2alpha; HIF-2-alpha; HIF2-alpha; HIF-related factor; HLF; HLF (HIF1alpha-like factor); HRF; hypoxia inducible factor 2 alpha; hypoxia inducible factor 2, alpha subunit; hypoxia inducible factor 2a; hypoxia inducible transcription factor 2alpha; hypoxia-inducible factor 2 alpha; hypoxia-inducible factor 2-alpha; Member of PAS protein 2; mHLF; MOP2; PAS domain-containing protein 2; PASD2; wu:fq36f01 |
| Gene Symbols | EPAS1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.