Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIF-2 alpha Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PIPA522696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522696 Supplier Invitrogen™ Supplier No. PA522696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: hypoxic lysates.

HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF-2 alpha
Applications ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate HRP
Formulation PBS with no preservative
Gene EPAS1
Gene Accession No. P97481, Q99814, Q9JHS1
Gene Alias basic-helix-loop-helix-PAS protein MOP2; bHLHe73; Class E basic helix-loop-helix protein 73; ECYT4; endothelial PAS domain protein 1; endothelial PAS domain protein 1b; endothelial PAS domain-containing protein 1; endothelial PAS domain-containing protein 1b; EPAS; Epas1; EPAS-1; epas1b; Hif like protein; HIF1 alpha-like factor; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF1alpha-like factor; HIF2 alpha; HIF-2 alpha; HIF2A; hif-2a; hif-2ab; HIF-2alpha; HIF-2-alpha; HIF2-alpha; HIF-related factor; HLF; HLF (HIF1alpha-like factor); HRF; hypoxia inducible factor 2 alpha; hypoxia inducible factor 2, alpha subunit; hypoxia inducible factor 2a; hypoxia inducible transcription factor 2alpha; hypoxia-inducible factor 2 alpha; hypoxia-inducible factor 2-alpha; Member of PAS protein 2; mHLF; MOP2; PAS domain-containing protein 2; PASD2; wu:fq36f01
Gene Symbols EPAS1
Host Species Rabbit
Immunogen Peptide derived from the C-terminus of mouse/human HIF-2 alpha protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100348809, 100771160, 13819, 2034, 29452, 443286
Target Species Human, Mouse, Rat, Rabbit, Monkey, Sheep, Hamster, Fish, Reptile
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.