Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF Prolyl Hydroxylases Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983815
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3815 Supplier Bioss Supplier No. BS15487R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Prolyl hydroxylase domain proteins HIF PHD1, HIF PHD2 and HIF PHD3 (known as PHD1, PHD2 and PHD3 in rodents, respectively) can hydroxylate HIF-alpha subunits. Hypoxia-inducible factor (HIF) is a transcriptional regulator important in several aspects of oxygen homeostasis. The prolyl hydroxylases catalyze the posttranslational formation of 4-hydroxyproline in HIF-alpha proteins. HIF PHD1, which is widely expressed, with highest levels of expression in testis, functions as a cellular oxygen sensor and is important in cell growth regulation. HIF PHD1 can localize to the nucleus or the cytoplasm and is also detected in hormone responsive tissues, such as normal and cancerous mammary, ovarian and prostate epithelium. HIF PHD1 is encoded by EGLN2, which maps to chromosome 19q13. 3.

Specifications

Antigen HIF Prolyl Hydroxylases
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EGLN2
Gene Accession No. Q96KS0
Gene Alias 0610011A13Rik; C85656; Egl nine homolog 2; egl-9 family hypoxia inducible factor 2; egl-9 family hypoxia-inducible factor 2; EGLN2; EIT6; EIT-6; Estrogen-induced tag 6; Falkor; HIF-1alpha prolyl-4-hydroxylase-1; HIF-P4H 1; Hif-p4h-1; HIFPH1; HIF-PH1; HIF-prolyl hydroxylase 1; HPH-1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 1; Ier4; immediate early response 4; P4H1; PHD1; PHD-1; Prolyl hydroxylase domain-containing protein 1; SM-20
Gene Symbols EGLN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HIF Prolyl Hydroxylases.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112398
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.