Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIF1A Monoclonal Antibody (H1alpha 67-7)
GREENER_CHOICE

Catalog No. AMPA6517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPA6517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPA6517 Supplier Invitrogen™ Supplier No. MA116517
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In Western blot, multiple bands may be seen at 120 kDa representing post-translational modification of HIF-1 alpha. Nuclear extracts are recommended. Suggested positive control: Cos7 CoCl2-treated nuclear extract.

HIF1-alpha (HIF1A) is a subunit of HIF1, which is a transcription factor found in mammalian cells cultured under reduced oxygen tension. HIF-1 is a heterodimer consisting of an alpha and beta subunit, both belonging to the basic-helix-loop-helix Per-aryl hydrocarbon receptor nuclear translocator-Sim (PAS) family of transcription factors. HIF1 functions as a transcriptional regulator of the adaptive response to hypoxia. Under hypoxic conditions, HIF-1 activates the transcription of over 40 genes, including erythropoietin, glucose transporters, glycolytic enzymes, vascular endothelial growth factor, HILPDA, and other genes whose protein products increase oxygen delivery or facilitate metabolic adaptation to hypoxia. HIF1-alpha regulates hypoxia-mediated apoptosis, cell proliferation and tumor angiogenesis. Hypoxia which induces p53 protein accumulation, directly interacts with HIF1-alpha and reduces hypoxia-induced expression of HIF1-alpha by promoting MDM2-mediated ubiquitination and proteasomal degradation under hypoxic conditions. Recent studies suggest that induction of NOX4 by HIF1-alpha contributes to maintain ROS levels after hypoxia and hypoxia-induced proliferation. In humans, it is located on the q arm of chromosome 14. The C-terminal of HIF1A binds to p300. p300/CBP-HIF complexes participate in the induction of hypoxia-responsive genes, including VEGF. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF1A
Applications Western Blot
Classification Monoclonal
Clone H1alpha 67-7
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene HIF1A
Gene Accession No. Q16665
Gene Alias AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L homeolog; hypoxia-inducible factor 1-alpha; hypoxia-inducible factor-1 alpha; hypoxia-inducible factor1alpha; member of PAS protein 1; member of PAS superfamily 1; MOP1; PAS domain-containing protein 8; PASD8; XELAEV_18039792mg; XELAEV_18041137mg; xhif1a
Gene Symbols HIF1A
Host Species Mouse
Immunogen Fusion protein containing amino acids 432-528 of human HIF-1 alpha.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3091
Target Species Human, Monkey
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.