Learn More
Invitrogen™ HIF1A Monoclonal Antibody (H1alpha67), DyLight™ 550

Description
In Western blot, multiple bands may be seen at 100-120 kDa representing post-translational modification of HIF-1 alpha. For WB, testing on nuclear extracts is recommended. Suggested positive control: Cos-7 nuclear extracts.
Specifications
Specifications
| Antigen | HIF1A |
| Applications | ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Proximity Ligation Assay, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | H1alpha67 |
| Concentration | 0.68 mg/mL |
| Conjugate | DyLight 550 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | HIF1A |
| Gene Accession No. | O35800, Q16665, Q61221, Q9I8A9, Q9XTA5 |
| Gene Alias | AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L homeolog; hypoxia-inducible factor 1-alpha; hypoxia-inducible factor-1 alpha; hypoxia-inducible factor1alpha; member of PAS protein 1; member of PAS superfamily 1; MOP1; PAS domain-containing protein 8; PASD8; XELAEV_18039792mg; XELAEV_18041137mg; xhif1a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.