Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF1A Mouse anti-Human, Unconjugated, Clone: HIF1A/84, Abnova™

Catalog No. 89337544 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-544 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-544 Supplier Abnova Corporation Supplier No. MAB21051
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

HIF1A Monoclonal antibody specifically detects HIF1A in Human samples. It is validated for IF, Flow Cyt.

Specifications

Antigen HIF1A
Applications Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone HIF1A/84
Conjugate Unconjugated
Description hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor)
Gene HIF1A
Gene Alias HIF-1alpha, HIF1, HIF1-ALPHA, MOP1, PASD8, bHLHe78
Host Species Mouse
Immunogen Recombinant protein corresponding to human HIF1A
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3091
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.