Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIF1AN Polyclonal Antibody
GREENER_CHOICE

Catalog No. APPA6593
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APPA6593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APPA6593 Supplier Invitrogen™ Supplier No. PA116593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: antigen standard for HIF1AN (transient overexpression lysate), rat astrocytes, C6 and U251 gliomas.

Hydroxylates HIF-1 alpha at 'Asn-803' in the C-terminal transactivation domain (CAD). Functions as an oxygen sensor and, under normoxic conditions, the hydroxylation prevents interaction of HIF-1 with transcriptional coactivators including Cbp/p300-interacting transactivator. Involved in transcriptional repression through interaction with HIF1A, VHL and histone deacetylases. Hydroxylates specific Asn residues within ankyrin repeat domains (ARD) of NFKB1, NFKBIA, NOTCH1, ASB4, PPP1R12A and several other ARD-containing proteins. Also hydroxylates Asp and His residues within ARDs of ANK1 and TNKS2, respectively. Negatively regulates NOTCH1 activity, accelerating myogenic differentiation. Positively regulates ASB4 activity, promoting vascular differentiation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF1AN
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Phosphate, tris citrate with 0.1% sodium azide; pH 7-8
Gene HIF1AN
Gene Accession No. Q8BLR9, Q9NWT6
Gene Alias 2310046M24Rik; A830014H24Rik; DKFZp762F1811; factor inhibiting HIF1; factor inhibiting HIF-1; FIH; FIH 1; FIH1; FIH-1; FLJ20615; FLJ22027; Hif1an; hypoxia inducible factor 1 alpha subunit inhibitor; hypoxia inducible factor 1 subunit alpha inhibitor; hypoxia inducible factor 1, alpha subunit inhibitor; hypoxia-inducible factor 1, alpha subunit inhibitor; hypoxia-inducible factor 1-alpha inhibitor; Hypoxia-inducible factor asparagine hydroxylase; peptide-aspartate beta-dioxygenase
Gene Symbols HIF1AN
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human Factor Inhibiting HIF-1 (between residues 300 and the C-terminus).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309434, 319594, 55662
Target Species Human, Mouse, Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.