Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF2-alpha Recombinant Rabbit Monoclonal Antibody (BL-95-1A2), Bethyl Laboratories

Catalog No. 501571260
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1260 20 μL
50-157-1259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1260 Supplier Bethyl Laboratories, Inc Supplier No. A700003T
Add to Cart
Add to Cart

Rabbit Recombit Monoclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.

Specifications

Antigen HIF2-alpha
Applications ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Recombinant Monoclonal
Clone BL-95-1A2
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene EPAS1
Gene Accession No. Q99814
Gene Alias Hypoxia inducible Factor 2 alpha, MOP2, Endothelial pas domain protein 1, Member of pas superfamily 2
Gene Symbols EPAS1
Host Species Rabbit
Immunogen Between 400 and 450
Purification Method Ion-exchange chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2034
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.