Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIF3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142580
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142580 Supplier Invitrogen™ Supplier No. PA5142580
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform a and c (NP_690007.1; NP_690008.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Cell growth and viability is compromised by oxygen deprivation (hypoxia). Hypoxia-inducible factors, including HIF-1 alpha, HIF-1 beta (also designated Arnt 1), EPAS-1 (also designated HIF-2 alpha) and HIF-3 alpha, induce glycolysis, erythropoiesis and angiogenesis in order to restore oxygen homeostasis. Hypoxia-inducible factors are members of the Per-Arnt-Sim (PAS) domain transcription factor family. In response to hypoxia, HIF-1 alpha is upregulated and forms a heterodimer with Arnt 1 to form the HIF-1 complex. The HIF-1 complex recognizes and binds to the hypoxia responsive element (HRE) of hypoxia-inducible genes, thereby activating transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF3A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene HIF3A
Gene Accession No. Q9Y2N7
Gene Alias Basic-helix-loop-helix-PAS protein MOP7; BHLHE17; Class E basic helix-loop-helix protein 17; HIF-3 alpha; HIF3 alpha 1; HIF3A; HIF-3A; HIF-3-alpha; HIF3-alpha; HIF3-alpha-1; hypoxia inducible factor 3 alpha; hypoxia inducible factor 3 alpha subunit; hypoxia inducible factor 3, alpha subunit; hypoxia inducible factor 3a; hypoxia inducible factor three alpha; hypoxia-inducible factor 3 alpha; hypoxia-inducible factor 3-alpha; Inhibitory PAS domain protein; IPAS; Member of PAS protein 7; Mop7; Neonatal and embryonic PAS protein; NEPAS; PAS domain-containing protein 7; PASD7
Gene Symbols HIF3A
Host Species Goat
Immunogen Peptide with sequence QDHRSTTELRKEKS-C (aa 3-17).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64344
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.